Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLIMP1/PRDM1 Antibody (646702), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB120447 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120447 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120447 Supplier Novus Biologicals Supplier No. FAB36081I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BLIMP1/PRDM1 Monoclonal specifically detects BLIMP1/PRDM1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen BLIMP1/PRDM1
Applications Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 646702
Conjugate Janelia Fluor 549
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias Beta-interferon gene positive regulatory domain I-binding factor, beta-interferon gene positive-regulatory domain I binding factor, BLIMP-1, BLIMP1MGC118925, blmp-1, B-lymphocyte-induced maturation protein 1, MGC118922, MGC118923, Positive regulatory domain I-binding factor 1, PR domain containing 1, with ZNF domain, PR domain zinc finger protein 1, PR domain-containing protein 1, PRDI-BF1MGC118924, PRDI-binding factor 1, PRDI-binding factor-1, PR-domain zinc finger protein 1
Host Species Mouse
Immunogen E. coli-derived recombinant human BLIMP1/PRDM1, Lys667-Cys789, Accession # O75626
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Cancer, Chromatin Research, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 639
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.