Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLM Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501556918
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
50-155-6918 100 μL
1 options

Catalog No. 50-155-6918

Supplier: Bethyl Laboratories, Inc A300110A

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The Bloom syndrome gene product is related to the RecQ subset of DExH box-containing DNA helicases and has both DNA-stimulated ATPase and ATP-dependent DNA helicase activities. Mutations causing Bloom syndrome delete or alter helicase motifs and may disable the 3′-5′ helicase activity. The normal protein may act to suppress inappropriate recombination.

Specifications

Antigen BLM
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene BLM
Gene Accession No. O88700, P54132
Gene Alias BLM
Gene Symbols BLM
Host Species Rabbit
Immunogen Between 1375 and C-term
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12144, 641
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.