Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLMH/Bleomycin Hydrolase Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB053432 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB053432 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB053432 Supplier Novus Biologicals Supplier No. NBP298334AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BLMH/Bleomycin Hydrolase Polyclonal specifically detects BLMH/Bleomycin Hydrolase in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen BLMH/Bleomycin Hydrolase
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias BHBMHBLM hydrolase, bleomycin hydrolase, EC 3.4.22.40
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant BLMH/Bleomycin Hydrolase (NP_848760.1; Asn 2-Glu 455).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents
Primary or Secondary Primary
Gene ID (Entrez) 642
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.