Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLNK Antibody (125), PerCP, Novus Biologicals™
SDP

Catalog No. NB316835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316835 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316835 Supplier Novus Biologicals Supplier No. NBP290140PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BLNK Monoclonal antibody specifically detects BLNK in Human samples. It is validated for ELISA

Specifications

Antigen BLNK
Applications ELISA
Classification Monoclonal
Clone 125
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias AGM4, B cell linker protein, BASH, B-cell adapter containing a SH2 domain protein, B-cell adapter containing a Src homology 2 domain protein, B-cell linker, B-cell linker protein, BLNK-s, Cytoplasmic adapter protein, LY57, MGC111051, SLP-65BLNK-S, SLP65Ly57, Src homology 2 domain-containing leukocyte protein of 65 kDa
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human BLNK (Accession#: AAH18906.1; Met 1-Ser 456).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29760
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.