Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLNK Antibody (OTI4C9), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N271855J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271855J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271855J646 Supplier Novus Biologicals Supplier No. NBP271855JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BLNK Monoclonal specifically detects BLNK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen BLNK
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4C9
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AGM4, B cell linker protein, BASH, B-cell adapter containing a SH2 domain protein, B-cell adapter containing a Src homology 2 domain protein, B-cell linker, B-cell linker protein, BLNK-s, Cytoplasmic adapter protein, LY57, MGC111051, SLP-65BLNK-S, SLP65Ly57, Src homology 2 domain-containing leukocyte protein of 65 kDa
Gene Symbols BLNK
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-292 of human BLNK (NP_037446) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29760.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.