Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLOC1S2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB329329 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329329 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329329 Supplier Novus Biologicals Supplier No. NBP300218APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BLOC1S2 Polyclonal antibody specifically detects BLOC1S2 in Human samples. It is validated for ELISA

Specifications

Antigen BLOC1S2
Applications ELISA
Classification Polyclonal
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias biogenesis of lysosomal organelles complex-1, subunit 2, BLOC-1 subunit 2, CEAP, centrosomal 10 kDa protein, centrosome protein oncogene, Centrosome-associated protein, subunit 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human BLOC1S2 (Uniprot#: Q6QNY1-2; Met1-Arg99).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 282991
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.