Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Blocker™ BSA

Catalog No. p-7229313
Click to view available options
Quantity:
125 mL
200 mL
3 x 200 mL
Buffer:
PBS
TBS

Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.

Blocker BSA is a 10% (w/v) solution of high-quality BSA that is useful for saturating excess protein-binding sites on membranes and microplates in immunoassays. It is most frequently diluted 10-fold (to 1% BSA) in 1X PBS or 1X TBS for initial testing. Blocker BSA is usually more effective than nonfat milk for biotin-avidin systems because it contains a single purified protein that is devoid of endogenous biotin.

TRUSTED_SUSTAINABILITY

Specifications

Description Blocker BSA in PBS (10X)
Product Type Blocker BSA
Quantity 3 x 200 mL
Form Liquid
For Use With (Application) Western Blot, ELISA, Immunohistochemistry (IHC)
Product Line Blocker™
Concentration 10% BSA
Buffer PBS
Blocking Agent Single purified protein
Content And Storage Store at 4°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.