Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Blocker™ Casein

Catalog No. p-9651005
Click to view available options
Quantity:
100 mL
1 L
3 x 1 L
Buffer:
PBS
TBS

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods.

Blocker Casein is a ready-to-use solution of purified casein for blocking steps in western blot, ELISA, IHC, and nucleic acid detection methods. Blocker Casein is 1% (w/v) casein, which corresponds to the optimal concentration for most applications.

Features of Blocker Casein include:
Purified casein—single-protein blocking buffer provides fewer chances of cross-reaction with assay components than serum or milk solutions
Easy to use—1% casein solution is ready to use; can be diluted further as needed
Safe—stable, thimerosal-free formulation

TRUSTED_SUSTAINABILITY

Specifications

Product Type Blocker Casein
Quantity 100 mL
Form Liquid
For Use With (Application) Western Blot, ELISA
Product Line Blocker™
Concentration 1X
Buffer PBS
Blocking Agent Single purified protein
Content And Storage Store at 4°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.