Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blooms Syndrome Protein Blm Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB100214IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100214IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100214IR Supplier Novus Biologicals Supplier No. NB100214IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Blooms Syndrome Protein Blm Polyclonal specifically detects Blooms Syndrome Protein Blm in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen Blooms Syndrome Protein Blm
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Bloom syndrome, Bloom syndrome, RecQ helicase-like, BSBloom syndrome protein, DNA helicase, RecQ-like type 2, EC 3.6.1, EC 3.6.4.12, MGC126616, MGC131618, RecQ protein-like 3, RecQ2, RECQ2MGC131620, RECQL3RECQL2
Gene Symbols BLM
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1425 and the C-terminus (residue 1477) of human Bloom Syndrome using the numbering given in entry NP_000048.1 (GeneID 641).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Genes Sensitive to DNA Damaging Agents, Phospho Specific, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 641
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.