Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blooms Syndrome Protein Blm Antibody, Novus Biologicals™
SDP

Catalog No. NB1001590 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001590 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001590 Supplier Novus Biologicals Supplier No. NB1001590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Blooms Syndrome Protein Blm Polyclonal specifically detects Blooms Syndrome Protein Blm in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen Blooms Syndrome Protein Blm
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 1-4 ug/mg of lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. O88700
Gene Alias Bloom syndrome, Bloom syndrome, RecQ helicase-like, BSBloom syndrome protein, DNA helicase, RecQ-like type 2, EC 3.6.1, EC 3.6.4.12, MGC126616, MGC131618, RecQ protein-like 3, RecQ2, RECQ2MGC131620, RECQL3RECQL2
Gene Symbols BLM
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1375 and the C-terminus (residue 1416) of mouse Bloom Syndrome Homolog using the numbering given in entry NP_031576.2 (GeneID 12144).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Genes Sensitive to DNA Damaging Agents, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 641
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.