Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nasco Blue Phantom Ultrasound Training Block

Shop All Simulaids, Inc. Products


Click to view available options
For Use With (Application):
Branched 2 Vessel
Branched 4 Vessel
Pediatric 4 Vessel
Regional Anesthesia
Soft Tissue Biopsy
Description
- Ultrasound training block
- Blue Phantom Series
Specifications
Specifications
| Product Type | Ultrasound Training Block |
| For Use With (Application) | Pediatric 4 Vessel |
| Color | Blue |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction