Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SI Analytics™ BlueLine Precision Electrode with Liquid Electrolyte

Catalog No. 15177872 Shop All Yellow Springs Instrument (YSI) Products
Click to view available options
Model:
pH 11
pH 14

Universal, precise pH measurement for the laboratory

  • Available with or without built-in NTC temperature sensors
  • For use with pH measurements in laboratory applications
  • Suitable for difficult samples
  • Optimum electrolyte flow without clogging by unique platinum wire junction
  • Different connection possibilities for a wide variety of pH meters
  • High performance membrane glass provides the best possible results
  • Easy to clean glass shaft
TRUSTED_SUSTAINABILITY

Specifications

Description Without Temperature Sensor
Connector Types S7 Screw Head
Junction Platinum Diaphragm
Material Glass
pH Range 0 to 14
Type Combination Electrode
Filling Solution Type 3 M KCl (Liquid)
For Use With (Application) pH Measurements in Laboratory Applications
For Use With (Equipment) Cable LB1A (Sold Separately)
Internal Reference Ag/AgCl
Membrane Type A Glass
Model pH 11
Temperature (Metric) Range -5°C to +100°C
Tip Type Conical
Length (Metric) 120 mm
Length (Metric) Cable Cable Sold Separately
Diameter (Metric) 12 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.