Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMAL1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NB100129

 View more versions of this product

Catalog No. NB100129


Description

Description

BMAL1 Polyclonal specifically detects BMAL1 in Human samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation.
Specifications

Specifications

BMAL1
Polyclonal
Western Blot 1:100 - 1:200, Simple Western 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
O00327
ARNTL
Recombinant protein corresponding to residues 294-624 of human BMAL1. [UniProt# O00327]
Unpurified
RUO
Primary
Human
Antisera
Western Blot, ChIP Assay
Unconjugated
Whole antisera with 0.02% Sodium Azide
aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, Member of PAS protein 3, member of PAS superfamily 3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3MGC47515
Rabbit
73 kDa
0.4 mL
Cholesterol Metabolism, Chromatin Research, Circadian Rhythm, Lipid and Metabolism, Neuroscience, Transcription Factors and Regulators
406
Store at 4C. Do not freeze.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only