Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMAL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90
Specifications
Antigen | BMAL1 |
---|---|
Dilution | Western Blot 1:100 - 1:200, Simple Western 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
Applications | Western Blot, ChIP Assay |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100129SS
![]() |
Novus Biologicals
NB100129SS |
0.1 mL |
Each for $181.90
|
|
|||||
NB100129
![]() |
Novus Biologicals
NB100129 |
0.4 mL | N/A | N/A | N/A | ||||
Description
BMAL1 Polyclonal specifically detects BMAL1 in Human samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation.Specifications
BMAL1 | |
Western Blot, ChIP Assay | |
Unconjugated | |
Rabbit | |
Cholesterol Metabolism, Chromatin Research, Circadian Rhythm, Lipid and Metabolism, Neuroscience, Transcription Factors and Regulators | |
Whole antisera with 0.02% Sodium Azide | |
aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, Member of PAS protein 3, member of PAS superfamily 3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3MGC47515 | |
ARNTL | |
IgG | |
Unpurified | |
73 kDa |
Western Blot 1:100 - 1:200, Simple Western 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
O00327 | |
406 | |
Recombinant protein corresponding to residues 294-624 of human BMAL1. [UniProt# O00327] | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title