Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMAL1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB1002288Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002288Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002288Z Supplier Novus Biologicals Supplier No. NB1002288AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BMAL1 Polyclonal specifically detects BMAL1 in Amphibian, Human, Mouse, Primate, Rat samples. It is validated for ChIP Assay, Flow Cytometry, Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen BMAL1
Applications ChIP Assay, Flow Cytometry, Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. O00327
Gene Alias aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, Member of PAS protein 3, member of PAS superfamily 3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3MGC47515
Gene Symbols ARNTL
Host Species Rabbit
Immunogen Bacterially expressed human BMAL1 (C-terminus). [UniProt# O00327].
Molecular Weight of Antigen 70 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Chromatin Research, Circadian Rhythm, Hypoxia, Lipid and Metabolism, Neuroscience, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 406
Target Species Amphibian, Human, Rat, Mouse, Primate, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.