Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMAL1 Antibody (OTI1H6), HRP, Novus Biologicals™
SDP

Catalog No. NBP270208H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270208H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270208H Supplier Novus Biologicals Supplier No. NBP270208H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BMAL1 Monoclonal specifically detects BMAL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen BMAL1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1H6
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias aryl hydrocarbon receptor nuclear translocator-like, aryl hydrocarbon receptor nuclear translocator-like protein 1, basic-helix-loop-helix-PAS orphan MOP3, BHLHE5, bHLHe5brain and muscle, bHLH-PAS protein JAP3, BMAL1TIC, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, Member of PAS protein 3, member of PAS superfamily 3, MOP3BMAL1c, PAS domain-containing protein 3, PASD3MGC47515
Gene Symbols ARNTL
Host Species Mouse
Immunogen Full length recombinant protein of human BMAL1 (NP_001169) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cholesterol Metabolism, Chromatin Research, Circadian Rhythm, Hypoxia, Lipid and Metabolism, Neuroscience, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 406.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.