Learn More
BMAL1 Polyclonal Antibody, Rockland™

Goat Polyclonal Antibody
$178.35 - $649.44
Specifications
| Antigen | BMAL1 |
|---|---|
| Concentration | 0.92 mg/mL |
| Applications | ELISA, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1591
![]()
|
Rockland Immunochemicals
600101ML8 |
100 μg |
Each for $649.44
|
|
|||||
50-199-1592
![]()
|
Rockland Immunochemicals
600101ML8S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human Bmal1. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to mouse, rat, golden hamster, horse, gallus gallus, Pongo abelii, Tyto alba, and Nannospalax galili.
The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene.Specifications
| BMAL1 | |
| ELISA, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Goat | |
| Human, Mouse | |
| O00327, Q9WTL8 | |
| 11865, 406 | |
| Primary | |
| Antibody |
| 0.92 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| Arnt3; Arntl; ARNT-like protein 1, brain and muscle; aryl hydrocarbon receptor nuclear translocator like; aryl hydrocarbon receptor nuclear translocator-like; Aryl hydrocarbon receptor nuclear translocator-like protein 1; basic-helix-loop-helix-PAS orphan MOP3; Basic-helix-loop-helix-PAS protein MOP3; BHLHE5; bHLH-PAS protein JAP3; Bmal1; Bmal1 protein; BMAL1b; bmal1b'; BMAL1c; BOS_14527; Brain and muscle ARNT-like 1; brain and muscle ARNT-like protein 1; CG8727 PA; Class E basic helix-loop-helix protein 5; cycle; JAP3; Member of PAS protein 3; member of PAS superfamily 3; MGC47515; MOP3; PAS domain-containing protein 3; PASD3; testis tissue sperm-binding protein Li 50e; Tic | |
| Arntl | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Arntl |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.