Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bmf Antibody (17A9), FITC, Novus Biologicals™
SDP

Catalog No. NB173245 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173245 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173245 Supplier Novus Biologicals Supplier No. NBP280091F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Bmf Monoclonal antibody specifically detects Bmf in Mouse, Rat samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Bmf
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 17A9
Conjugate FITC
Formulation PBS
Gene Alias Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065
Host Species Rat
Immunogen Recombinant full length mouse Bmf and synthetic peptides corresponding to aa 13-27 and aa 112-126 of mouse Bmf.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 90427
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.