Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BMH 21
GREENER_CHOICE

Catalog No. p-7161607
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

RNA polymerase I inhibitor; also p53 pathway activator

BMH 21 is a p53 pathway activator; induces expression of p53 target genes. Also RNA polymerase 1 inhibitor; promotes degradation of RPA194. Reduces viability of multiple tumor cell lines and attenuates tumor growth of A375 melanoma cell xenografts in mice.

Specifications

Chemical Name or Material N-[2-(Dimethylamino)ethyl]-12-oxo-12 H-benzo[g]pyrido[2,1-b]quinazoline-4-carboxamide
CAS 896705-16-1
Quantity 10 mg
Target RNA/DNA Polymerase Inhibitors
Molecular Formula C21H20N4O2
Formula Weight 360.41
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.