Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMI-1 Antibody (116), DyLight 594
SDP

Catalog No. NB087799 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087799 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087799 Supplier Novus Biologicals Supplier No. NBP289213DL594

Rabbit Monoclonal Antibody

BMI-1 Monoclonal specifically detects BMI-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen BMI-1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 116
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias B lymphoma Mo-MLV insertion region 1 homolog, B lymphoma Mo-MLV insertion region 1 homolog (mouse), BMI1 polycomb ring finger oncogene, FLVI2/BMI1, MGC12685, murine leukemia viral (bmi-1) oncogene homolog, PCGF4, polycomb complex protein BMI-1, polycomb group protein Bmi1, polycomb group ring finger 4, RING finger protein 51, RNF51Polycomb group RING finger protein 4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human BMI-1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cancer Stem Cells, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, Hematopoietic Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 648
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.