Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMI-1 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB11040823R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040823R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11040823R Supplier Novus Biologicals Supplier No. NB11040823R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BMI-1 Polyclonal specifically detects BMI-1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BMI-1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P35226
Gene Alias B lymphoma Mo-MLV insertion region 1 homolog, B lymphoma Mo-MLV insertion region 1 homolog (mouse), BMI1 polycomb ring finger oncogene, FLVI2/BMI1, MGC12685, murine leukemia viral (bmi-1) oncogene homolog, PCGF4, polycomb complex protein BMI-1, polycomb group protein Bmi1, polycomb group ring finger 4, RING finger protein 51, RNF51Polycomb group RING finger protein 4
Gene Symbols BMI1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region (within residues 1-100) of human Bmi1. [Swiss-Prot# P35226]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Hematopoietic Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 648
Test Specificity Human and mouse. Immunogen sequence has 92% homology to primate, feline, chicken, bovine, rabbit, rat and Zebrafish protein.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.