Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMI-1 Rabbit anti-Human, Rat, Clone: 6H9B0, Novus Biologicals™
SDP

Catalog No. NB166220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166220 20 μg
NB166219 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166220 Supplier Novus Biologicals Supplier No. NBP31556220UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BMI-1 Monoclonal antibody specifically detects BMI-1 in Human, Rat samples. It is validated for Western Blot

Specifications

Antigen BMI-1
Applications Western Blot
Classification Monoclonal
Clone 6H9B0
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias B lymphoma Mo-MLV insertion region 1 homolog, B lymphoma Mo-MLV insertion region 1 homolog (mouse), BMI1 polycomb ring finger oncogene, FLVI2/BMI1, MGC12685, murine leukemia viral (bmi-1) oncogene homolog, PCGF4, polycomb complex protein BMI-1, polycomb group protein Bmi1, polycomb group ring finger 4, RING finger protein 51, RNF51Polycomb group RING finger protein 4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 227-326 of human BMI-1 (P35226).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cancer Stem Cells, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, Hematopoietic Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 648
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.