Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-1/PCP Antibody (OTI3E9), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP270255G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270255G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270255G Supplier Novus Biologicals Supplier No. NBP270255G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BMP-1/PCP Monoclonal specifically detects BMP-1/PCP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BMP-1/PCP
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3E9
Conjugate DyLight 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BMP-1, bone morphogenetic protein 1, EC 3.4.24, EC 3.4.24.19, FLJ44432, Mammalian tolloid protein, mTld, PCOLC, PCP, PCP2, procollagen C-endopeptidase, Procollagen C-proteinase, TLD
Gene Symbols BMP1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 225-617 of human BMP-1/PCP (NP_001190) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 649.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.