Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-15/GDF-9B Antibody (BMP15/4263), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB389850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389850 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389850 Supplier Novus Biologicals Supplier No. NBP308876IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BMP-15/GDF-9B Monoclonal antibody specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen BMP-15/GDF-9B
Applications Western Blot, ELISA
Classification Monoclonal
Clone BMP15/4263
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4
Host Species Mouse
Immunogen Recognizes sequence SAEVTASSSKHSGPENNQC on the C-terminus of human BMP-15/GDF-9B (Uniprot: O95972)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Immunology, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 9210
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.