Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-15/GDF-9B Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB164503 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164503 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164503 Supplier Novus Biologicals Supplier No. NBP281743H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BMP-15/GDF-9B Polyclonal antibody specifically detects BMP-15/GDF-9B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen BMP-15/GDF-9B
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias BMP-15, bone morphogenetic protein 15, GDF-9B, Growth/differentiation factor 9B, ODG2, POF4
Host Species Rabbit
Immunogen Rabbit polyclonal BMP15 antibody was raised against an 18 amino acid peptide near the amino terminus of human BMP15. The immunogen is located within amino acids 60 - 110 of BMP15. Amino Acid Squence: LYRRSADSHGHPRENRTI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Immunology, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 9210
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.