Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-2 Antibody (110), PE, Novus Biologicals™
SDP

Catalog No. NB307415 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307415 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307415 Supplier Novus Biologicals Supplier No. NBP289490PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BMP-2 Monoclonal antibody specifically detects BMP-2 in Human, Mouse, Rat, Canine, Macaque samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen BMP-2
Applications Immunocytochemistry
Classification Monoclonal
Clone 110
Conjugate PE
Dilution Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human/Mouse/Rat/Rhesus/Canine BMP-2 (Accession#: NP_001191.1; Gln283-Arg396).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Neuronal Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 650
Target Species Human, Mouse, Rat, Canine, Macaque
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.