Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-4 Antibody (OTI6B7), Novus Biologicals™
SDP

Catalog No. NBP147666 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147666 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147666 Supplier Novus Biologicals Supplier No. NBP147666
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BMP-4 Monoclonal specifically detects BMP-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BMP-4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI6B7
Concentration 0.42 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100000, Immunocytochemistry/Immunofluorescence 1:100
Gene Alias BMP-2B, BMP2B1, BMP2BMCOPS6, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, OFC11, ZYME
Gene Symbols BMP4
Host Species Mouse
Immunogen Recombinant fragment expressed in E.coli corresponding to amino acids 293-408 of human BMP4
Molecular Weight of Antigen 46.4 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Embryonic Stem Cell Markers, Stem Cell Markers, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 652
Test Specificity This antibody is specific for Homo sapiens bone morphogenetic protein 4 (BMP4), transcript variant 2.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.