Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-6 Antibody (74219.11), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1201089A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1201089A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1201089A70 Supplier Novus Biologicals Supplier No. NB12010859AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BMP-6 Monoclonal specifically detects BMP-6 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen BMP-6
Applications Western Blot, ELISA
Classification Monoclonal
Clone 74219.11
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, VGR1, VGR-1
Gene Symbols BMP6
Host Species Mouse
Immunogen Recombinant full length protein (Human) expressed in CHO cells.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 654.0
Test Specificity This shows less than 5 % cross-reactivity with recombinant human BMP5 and BMP7 and no cross-reactivity with recombinant human BMP2 and BMP4 under reducing conditions.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.