Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ BMP™61 BMP™71 Series ToughBond™ Ultra Aggressive Polyester Label Supply - White
Click to view available options
Color:
White
Description
- Glossy white polyester with permanent rubber based adhesive is thermal transfer printable
- Material is designed for high adhesion to textured metals and low surface energy plastics
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
- UL approved
- CSA accepted
- Service temperature of -40 to 248° F (-40 to 120° C)
Specifications
Specifications
Color | White |
For Use With (Equipment) | BMP™61 Labeling System, BMP™71 Label Printer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction