Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP-8b Antibody (13E6) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP242693 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242693 Supplier Novus Biologicals Supplier No. NBP242693
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, ELISA

BMP-8b Monoclonal specifically detects BMP-8b in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BMP-8b
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 13000000
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Formulation PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Gene Accession No. NP_001711
Gene Alias BMP-8, bone morphogenetic protein 8b, MGC131757
Gene Symbols BMP8B
Host Species Mouse
Immunogen Recombinant human BMP8b (264-402aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 656
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.