Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR-IA/ALK-3 Antibody (002), PE, Novus Biologicals™
SDP

Catalog No. NB333356 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333356 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333356 Supplier Novus Biologicals Supplier No. NBP290398PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BMPR-IA/ALK-3 Monoclonal antibody specifically detects BMPR-IA/ALK-3 in Mouse samples. It is validated for ELISA

Specifications

Antigen BMPR-IA/ALK-3
Applications ELISA
Classification Monoclonal
Clone 002
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias ACVRLK310q23del, ALK-3, ALK3EC 2.7.11.30, BMP type-1A receptor, BMPR-1A, bone morphogenetic protein receptor type-1A, bone morphogenetic protein receptor, type IA, CD292, CD292 antigen, EC 2.7.11, Serine/threonine-protein kinase receptor R5, SKR5, type II-like kinase 3
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse BMPR-IA/ALK-3 (Accession#: NP_033888.2, Met 1-Arg 152)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 657
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.