Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR-IB/ALK-6 Antibody [DyLight 405], Novus Biologicals™
SDP

Catalog No. AF505E Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF505E 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF505E Supplier Novus Biologicals Supplier No. AF505E
Add to Cart
Add to Cart

Goat Polyclonal Antibody

BMPR-IB/ALK-6 Polyclonal antibody specifically detects BMPR-IB/ALK-6 in Human samples. It is validated for Western Blot

Specifications

Antigen BMPR-IB/ALK-6
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias activin receptor-like kinase 6, ALK-6, ALK6CDw293, BMP type-1B receptor, BMPR-1B, bone morphogenetic protein receptor type-1B, bone morphogenetic protein receptor, type IB, CDw293 antigen, EC 2.7.11, EC 2.7.11.30, serine/threonine receptor kinase
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human BMPR-IB/ALK-6, Lys14-Arg126, Accession # O00238
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 658
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.