Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR-IB/ALK-6 Antibody (MM0055-3E12) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP212031 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212031 0.1 mg
NB019005 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP212031 Supplier Novus Biologicals Supplier No. NBP212031

Mouse Monoclonal Antibody

BMPR-IB/ALK-6 Monoclonal specifically detects BMPR-IB/ALK-6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen BMPR-IB/ALK-6
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0055-3E12
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. O00238
Gene Alias activin receptor-like kinase 6, ALK-6, ALK6CDw293, BMP type-1B receptor, BMPR-1B, bone morphogenetic protein receptor type-1B, bone morphogenetic protein receptor, type IB, CDw293 antigen, EC 2.7.11, EC 2.7.11.30, serine/threonine receptor kinase
Gene Symbols BMPR1B
Host Species Mouse
Immunogen Human recombinant BMPR-IB/ALK-6 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 658
Test Specificity In direct ELISAs and Western blots, 100% good cross-reactivity with recombinant mouse BMPR-IB, but no cross-reactivity with recombinant human and mouse BMPR-IA is observed.
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.