Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMPR-IB/ALK-6 Antibody (MM0055-3E12) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
| Antigen | BMPR-IB/ALK-6 |
|---|---|
| Clone | MM0055-3E12 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
Description
BMPR-IB/ALK-6 Monoclonal specifically detects BMPR-IB/ALK-6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| BMPR-IB/ALK-6 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| activin receptor-like kinase 6, ALK-6, ALK6CDw293, BMP type-1B receptor, BMPR-1B, bone morphogenetic protein receptor type-1B, bone morphogenetic protein receptor, type IB, CDw293 antigen, EC 2.7.11, EC 2.7.11.30, serine/threonine receptor kinase | |
| BMPR1B | |
| Protein G purified |
| MM0055-3E12 | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers | |
| O00238 | |
| 658 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title