Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR-II Antibody (MM0060-9A10), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB361389 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361389 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361389 Supplier Novus Biologicals Supplier No. NBP212064MFV450

Mouse Monoclonal Antibody

BMPR-II Monoclonal antibody specifically detects BMPR-II in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen BMPR-II
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0060-9A10
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias BMP type II receptor, BMP type-2 receptor, BMPR-2, BMPR3, BMPR-IIprimary pulmonary hypertension 1, Bone morphogenetic protein receptor type II, bone morphogenetic protein receptor type-2, bone morphogenetic protein receptor, type II (serine/threonine kinase), BRK-3, EC 2.7.11, EC 2.7.11.30, FLJ41585, FLJ76945, PPH1BMR2, T-ALK, type II activin receptor-like kinase, type II receptor for bone morphogenetic protein-4
Host Species Mouse
Immunogen Human recombinant BMPR-II (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 659
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.