Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR-II Antibody, Novus Biologicals™
SDP

Catalog No. NBP132218 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132218 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132218 Supplier Novus Biologicals Supplier No. NBP132218
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BMPR-II Polyclonal specifically detects BMPR-II in Human samples. It is validated for Western Blot.

Specifications

Antigen BMPR-II
Applications Western Blot
Classification Polyclonal
Concentration 0.89mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 1XPBS, 1% BSA, 10% Glycerol (pH7). with 0.025% Proclin 300
Gene Alias BMP type II receptor, BMP type-2 receptor, BMPR-2, BMPR3, BMPR-IIprimary pulmonary hypertension 1, Bone morphogenetic protein receptor type II, bone morphogenetic protein receptor type-2, bone morphogenetic protein receptor, type II (serine/threonine kinase), BRK-3, EC 2.7.11, EC 2.7.11.30, FLJ41585, FLJ76945, PPH1BMR2, T-ALK, type II activin receptor-like kinase, type II receptor for bone morphogenetic protein-4
Gene Symbols BMPR2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human BMPR2. The exact sequence is proprietary.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 659
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.