Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR-II Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094069 100 μL
NB094070 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094069 Supplier Novus Biologicals Supplier No. NBP303627100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BMPR-II Polyclonal specifically detects BMPR-II in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.

Specifications

Antigen BMPR-II
Applications Western Blot, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias BMP type II receptor, BMP type-2 receptor, BMPR-2, BMPR3, BMPR-IIprimary pulmonary hypertension 1, Bone morphogenetic protein receptor type II, bone morphogenetic protein receptor type-2, bone morphogenetic protein receptor, type II (serine/threonine kinase), BRK-3, EC 2.7.11, EC 2.7.11.30, FLJ41585, FLJ76945, PPH1BMR2, T-ALK, type II activin receptor-like kinase, type II receptor for bone morphogenetic protein-4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 27-150 of human BMPR-II (NP_001195.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Mesenchymal Stem Cell Markers, Protein Kinase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 659
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.