Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BMS 193885
GREENER_CHOICESDP

Catalog No. 324250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-425-0 50 mg
32-4210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-425-0 Supplier Tocris Bioscience™ Supplier No. 3242/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, competitive NPY Y1 receptor antagonist

Potent, competitive neuropeptide (NPY) Y1 antagonist (Ki = 3.3 nM, IC50 = 5.9 nM) that displays > 47, > 100, > 160, > 160 and > 160-fold selectivity over σ1, α1, Y2, Y4 and Y5 receptors respectively. Reduces food intake and body weight via central Y1 inhibition and is brain penetrant.

Chemical Identifiers

CAS 679839-66-8
Molecular Formula C36H48N4O9

Specifications

Quantity 50 mg
Formula Weight 680.79
Purity >99%
Molecular Weight (g/mol) 680.80
Chemical Name or Material BMS 193885
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.