Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BMS 299897
GREENER_CHOICESDP

Catalog No. 287010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-701-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 28-701-0 Supplier Tocris Bioscience™ Supplier No. 2870/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent γ-secretase inhibitor

Orally active, potent γ-secretase inhibitor (IC50 = 12 nM). Inhibits Aβ40 and Aβ42 formation in vitro (IC50 values are 7.4 and 7.9 nM respectively) and reduces Aβ in the brain, plasma and cerebrospinal fluid in vivo. Exhibits no Notch toxicity. Brain penetrant.

Chemical Identifiers

CAS 290315-45-6
Molecular Formula C24H21ClF3NO4S

Specifications

Quantity 10 mg
Formula Weight 511.94
Purity >98%
Molecular Weight (g/mol) 511.94
Chemical Name or Material BMS 299897
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.