Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ BMS 303141

Click to view available options
Quantity:
10 mg
50 mg
Description
ATP citrate lyase (ACL) inhibitor (IC50 = 0.13 μM for human recombinant ACL); blocks lipid synthesis (IC50 = 8 μM in HepG2 cells). Displays no cytotoxicity up to a concentration of 50 μM. Lowers plasma glucose and triglycerides in a mouse model of hyperlipidemia. Orally bioavailable.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 424.3 |
Purity | 0.98 |
Molecular Weight (g/mol) | 424.29 |
Chemical Name or Material | BMS 303141 |
Safety and Handling
Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction