Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ BMS 345541

Click to view available options
Quantity:
10 mg
50 mg
Description
BMS 345541 is a selective allosteric inhibitor of IKK (IC50 values are 0.3 and 4.0 μM for IKKβ and IKKα respectively). Exhibits no effect against a panel of 15 other kinases. Attenuates LPS-induced cytokine production in vitro and blocks NFκB dependent transcription in mice. Also suppresses joint destruction in a mouse model of arthritis.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | BMS 345541 |
| Purity | >99% |
| Molecular Weight (g/mol) | 291.78 |
| Inhibitors | BMS 345541 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction