Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BMS CCR2 22
GREENER_CHOICE

Catalog No. 312910
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-291-0 10 mg
31-291 1 mg
31-295-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 31-291-0 Supplier Tocris Bioscience™ Supplier No. 3129/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity, potent CCR2 antagonist

High affinity CCR2 chemokine receptor antagonist (IC50 = 5.1 nM). Displays potent functional antagonism, IC50 values are 1 and 18 nM for chemotaxis and antagonism of calcium flux respectively. Selective over CCR3.

Chemical Identifiers

CAS 445479-97-0
Molecular Formula C28H34F3N5O4S

Specifications

Quantity 10 mg
Formula Weight 593.66
Purity >98%
Molecular Weight (g/mol) 593.666
Chemical Name or Material BMS CCR2 22
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.