Learn More
Description
Specifications
Specifications
| Antigen | BNC1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | PBS & 2% Sucrose. with No Preservative |
| Gene Alias | basonuclin, basonuclin 1, BNC, BSN1, HsT19447, zinc finger protein basonuclin, zinc finger protein basonuclin-1 |
| Gene Symbols | BNC1 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptides corresponding to Bnc1 (basonuclin 1) The peptide sequence was selected from the C terminal of Bnc1. Peptide sequence ETSEDHFRAAYLLQDVAKEAYQDVAFTPQASQTSVIFKGTSGMGSLVYPI. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
