Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNC1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224721 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224721 0.1 mg
NBP224721SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224721 Supplier Novus Biologicals Supplier No. NBP224721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BNC1 Polyclonal specifically detects BNC1 in Human, Primate samples. It is validated for Western Blot.

Specifications

Antigen BNC1
Applications Western Blot, KnockDown
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 4-7 ug/ml, KnockDown Validated reported in scientific literature (PMID 34526665)
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q01954
Gene Alias basonuclin, basonuclin 1, BNC, BSN1, HsT19447, zinc finger protein basonuclin, zinc finger protein basonuclin-1
Gene Symbols BNC1
Host Species Rabbit
Immunogen A portion of amino acids 330-380 of human Bnc 1 protein was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 646
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey and dog, 91% homologous in horse, mouse and rat, and 83% homologous in cow and opossum.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.