Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNIP2 Antibody (OTI1C12), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP27028A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27028A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27028A70 Supplier Novus Biologicals Supplier No. NBP270258AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BNIP2 Monoclonal specifically detects BNIP2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen BNIP2
Applications Western Blot
Classification Monoclonal
Clone OTI1C12
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BCL2/adenovirus E1B 19 kDa protein-interacting protein 2, BCL2/adenovirus E1B 19kDa interacting protein 2, BCL2/adenovirus E1B 19kD-interacting protein 2, BNIP-2, NIP2
Gene Symbols BNIP2
Host Species Mouse
Immunogen Full length recombinant protein of human BNIP2 (NP_004321) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 663.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.