Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BNIP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$243.50 - $353.50
Specifications
Antigen | BNIP3 |
---|---|
Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:5000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056150T
![]() |
Novus Biologicals
NB10056150SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB10056150
![]() |
Novus Biologicals
NB10056150 |
0.05 mL |
Each for $353.50
|
|
|||||
Description
BNIP3 Polyclonal specifically detects BNIP3 in Human, Rat, Porcine, Chicken, Xenopus, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BNIP3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis, Autophagy, Cancer, Hypoxia, Tumor Suppressors | |
Whole antisera with 0.05% Sodium Azide | |
664 | |
A synthetic peptide corresponding to amino acids 122-139 (SRPENIPPKEFLFKHPKR) of human NIP3 was used aas immunogen; GenBank no. gi 7669481 ref NP_004043.2 . | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Rat, Pig, Chicken, Xenopus species, Mouse (Negative) | |
BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3 | |
BNIP3 | |
IgG | |
Unpurified | |
NIP3 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title