Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNIP3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056150 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056150 0.05 mL
NB10056150T 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056150 Supplier Novus Biologicals Supplier No. NB10056150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

BNIP3 Polyclonal specifically detects BNIP3 in Chicken, Human, Mouse (Negative), Pig, Rat, Xenopus species samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen BNIP3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:5000
Formulation Whole antisera with 0.05% Sodium Azide
Gene Alias BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3
Gene Symbols BNIP3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 122-139 (SRPENIPPKEFLFKHPKR) of human NIP3 was used aas immunogen; GenBank no. gi 7669481 ref NP_004043.2 .
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Hypoxia, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 664
Test Specificity NIP3
Target Species Chicken, Rat, Human, Mouse (Negative), Pig, Mouse (Negative), Rat, Xenopus species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.