Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNIP3 Antibody (ANa40), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB1001543Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001543Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001543Y Supplier Novus Biologicals Supplier No. NB1001543AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BNIP3 Monoclonal specifically detects BNIP3 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen BNIP3
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone ANa40
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Gene Accession No. Q12983
Gene Alias BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3
Gene Symbols BNIP3
Host Species Mouse
Immunogen Recombinant fragment (Residues 1-163 of Human BNIP3).
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Hypoxia, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 664
Test Specificity BNIP3 (ANa40)
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.