Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNIP3L Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP178264 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178264 0.1 mL
NBP178264SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP178264 Supplier Novus Biologicals Supplier No. NBP178264
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BNIP3L Polyclonal specifically detects BNIP3L in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen BNIP3L
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:300
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. O60238
Gene Alias Adenovirus E1B19K-binding protein B5, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like, BCL2/adenovirus E1B 19-kd protein-interacting protein 3a, BCL2/adenovirus E1B 19kDa interacting protein 3-like, BNIP3a, BNIP3H, NIP3L, NIP3-like protein X, Nix, NIXBCL2/adenovirus E1B 19kD-interacting protein 3-like
Gene Symbols BNIP3L
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human BNIP3L protein (between residues 50-100) [UniProt O60238]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 665
Test Specificity Human and mouse. Immunogen has 100% identity to bovine.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.