Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BNP Antibody (SR1505), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32176550UL
Description
BNP Monoclonal antibody specifically detects BNP in Human samples. It is validated for Western BlotSpecifications
BNP | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1505 | |
Western Blot 1:1000-1:5000 | |
BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein | |
A synthesized peptide derived from human BNP (Uniprot #: P16860) | |
50 μg | |
Biologically Active Proteins, Cardiovascular Biology, Cytokine Research, Immunology, Neuroscience, Neurotransmission | |
4879 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction